Masterflex® L/S® Precision Pump Tubing, Tygon® E-Lab, L/S 18; 50 ft
Supplier: Avantor Fluid Handling
VWR Item#MFLX06509-18
Your Organization's Price:
Ideal for general transfer applications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Tubing Size: L/S 18
<B>Applications</B>: General laboratory use<br /><B>Characteristics</B>: Nontoxic, nonaging, nonoxidizing, easy flow monitoring, transparent