Kjeldahl distillation apparatus
Supplier: witeg Labortechnik
2.530.000
WITG2.530.000EA 0 INR
WITG2.530.000
Kjeldahl distillation apparatus
Distillation Systems Kjeldahl Distillation Systems
Kjeldahl distillation unit made of borosilicate glass.
Ordering information: Includes condenser, funnel stopper (ST 10/19), dropping funnel (50 ml), Kjedahl flask (500 ml), adapters and fittings.
Order Now
SPECIFICATIONS
Environmentally Preferable