VWR®, Magnetic hotplate stirrers, standard series
Supplier: VWR Collection
97042-636
VWRU97042-636EA 0 INR
VWRU97042-636
VWR®, Magnetic hotplate stirrers, standard series
Stirrers Magnetic Hotplate Stirrers
These Standard series hotplate stirrers with microprocessor control were designed for general laboratory heating and stirring. These analogue units equipped with easy to use, dial-adjustment controls. An internal RTD accurately measures and controls temperature. Hot symbol warning light is illuminated when heat is turned on and remains on until top plate cools down.
  • Cool touch housing made of heat-resistant polymer
  • Speed ramping feature to prevent splashing, improve magnetic coupling, and provide excellent low end control
  • Built in support rod holder