VWR® Talon®, Clamps, Three Pronged
Supplier: VWR Collection
21570-125 12621-236 21570-126 21570-127 21570-302 12621-238 21570-404 21572-556 12621-240 12621-250 97035-502 97035-504
VWRU21570-125EA 0 INR
VWRU21570-125 VWRU12621-236 VWRU21570-126 VWRU21570-127 VWRU21570-302 VWRU12621-238 VWRU21570-404 VWRU21572-556 VWRU12621-240 VWRU12621-250 241-0404 241-0405 241-0406 VWRU97035-502 VWRU97035-504
VWR® Talon®, Clamps, Three Pronged
Clamps General Purpose Clamps
Stainless steel or nickel-plated zinc. These clamps securely hold every type of glassware and apparatus.
  • Extension arms allow placement of apparatus at various distances from lab frames without compromising the integrity of the experiment
  • Stainless steel clamps: Exceptional chemical resistance, clamps are electropolished and autoclavable
  • Nickel-plated zinc clamps: High tensile strength
  • Supplied with non-slip vinyl sleeves, and fiberglass covers for temperatures above 100 °C
Long, seamless tubing attaches clamp head securely and offers easy positioningMore Product Information