VWR®sympHony™, Multi-parameter meters, bench top
Supplier: VWR Collection
89231-664 89231-692 89231-698
VWRU89231-664EA 0 INR
VWRU89231-664 VWRU89231-692 VWRU89231-698
VWR®sympHony™, Multi-parameter meters, bench top
Electrochemistry Meters pH and Multi-Parameter Meters
These bench top meters are specially designed for use in environmental field applications and in the laboratory. The meters are rugged, durable and versatile enough to fit for unique testing needs.
  • Simple to operate and maintain
  • Consistently accurate and reliable measurements
  • Configurable to each users unique needs
  • Meet IP54 specifications for protection from water
These meters are simple to operate and offer an intuitive user interface allowing for easy set up and use from day one. Step-by-step on screen prompts enable both new and seasoned users toMore Product Information