VWR®, Labelling Tapes, General Purpose Laboratory
Supplier: VWR Collection
89098-062 89097-996 89098-004 89097-998 89097-990 89097-986 89098-002 89098-000 89097-920 89098-058 89098-072 89098-094 89097-956 89097-958 89097-962 89097-964 89098-032 89098-028 89098-024 89098-026 89098-030 89098-034 89098-036
VWRU89098-062EA 0 INR
VWRU89098-062 VWRU89097-996 VWRU89098-004 VWRU89097-998 VWRU89097-990 VWRU89097-986 VWRU89098-002 VWRU89098-000 VWRU89097-920 VWRU89098-058 VWRU89098-072 VWRU89098-094 VWRU89097-956 VWRU89097-958 VWRU89097-962 VWRU89097-964 VWRU89098-032 VWRU89098-028 VWRU89098-024 VWRU89098-026 VWRU89098-030 VWRU89098-034 VWRU89098-036
VWR®, Labelling Tapes, General Purpose Laboratory
Tapes
VWR® general-purpose laboratory labelling tape sticks to virtually any surface, can be marked with most writing utensils, and reliably withstands extreme temperature variations. It can be written on with pencil, fiber-tip or ball point pen, and lab markers.
  • Vinyl-coated soft satin-finish paper
  • Adhesive sticks to virtually any surface
  • Tape removal leaves no residue
This durable self-adhesive tape is sutiable for general laboratory use and is excellent for applications in refrigeration. It can withstand temperatures from –23 to 121 °C (–10 to 250 °F)More Product Information