VWR®, Tubing, Silicone
Supplier: VWR Collection
89068-432 89068-482 89068-436 89068-478
VWRU89068-432EA 0 INR
VWRU89068-432 VWRU89068-482 VWRU89068-436 VWRU89068-478
VWR®, Tubing, Silicone
Tubing Transfer Tubing
Platinum-cured silicone tubing is flexible, durable, low volatile grade, and translucent. This tubing is ideal for a variety of transfer applications such as pharmaceutical, laboratory, food and beverage, and bioprocess manufacturing. It features an ultra-smooth interior surface that provides uniform fluid flow and resistance to bacteria build-up.
  • Can be sterilised by autoclaving and gamma-irradiation
  • Imparts no taste or odour to critical stream
Certifications: Meets USP Class VI; USP 661 (Physicochemical Tests); ISO10993, EP 3.1.9, 3-A standards andMore Product Information