VWR®, Undercounter Laboratory Refrigerator
Supplier: VWR Collection
89031-974
VWRU89031-974EA 0 INR
VWRU89031-974
VWR®, Undercounter Laboratory Refrigerator
Refrigerators
This laboratory refrigerator unit is designed to meet the demanding standards of clinical, life science, pharmaceutical, biotechnology and industrial applications.
  • Exceptional temperature control and uniformity
  • Microprocessor control with user-programmable functions and features
  • Built-in front-breather condenser
Units offer exceptional temperature control and uniformity via a door-mounted microprocessor controller and interior forced air circulation. The easy-to-read controller features an upward angled LED display and keypad. Digital temperatureMore Product Information