VWR® Jaw Style Tubing Clamp, Acetal
Supplier: VWR Collection
70690-116
VWRU70690-116EA 0 INR
VWRU70690-116
VWR® Jaw Style Tubing Clamp, Acetal
Clamps Tubing and Sanitary Clamps
Tube clamp is molded of strong, durable plastic with serrated jaws 270 mm (1¹⁄₁₆") long.
  • Designed for one-handed operation
  • Strong, durable plastic construction
  • Serrated jaws
  • Autoclavable
Attach at any point on tubing without disconnecting tubing
When not in use, the circular opening (approximately 60 mm (¼") diameter) at hinge end allows clamp to remain attached to tubing having 6 to 100 mm (¼ to ⅜") OD with little to no restriction in flow. Overall length is 590 mm (2⁵⁄₁₆"). Autoclavable.
Designed to be operated with one hand, the clamp locksMore Product Information