VWR® Traceable®, Infrared thermometer
Supplier: VWR Collection
36934-182
VWRU36934-182EA 0 INR
VWRU36934-182
VWR® Traceable®, Infrared thermometer
Thermometers Digital Thermometers
This thermometer uses a laser-sighting beam to aim precisely at target surfaces.
  • One-handed point and shoot readings.
  • Instant, less-than-a-second temperature readings of any surface, °F/°C switchable
  • Features minimum/maximum memory, average readings and high/low audible alarms
Ideal for food preparation, pharmaceuticals, field use, electronics and safety inspection.
Certifications: Compliance with ISO/IEC 17025:2005 (1750.01); accredited by A2LA indicating traceability to standards provided by NIST.
Delivery information: Supplied with battery,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable