VWR® Traceable® Ultra, Digital Thermometers

Supplier: VWR Collection
23226-658 23609-176
VWRU23226-658EA 0 INR
VWRU23226-658 620-2081 620-2079 620-2080 VWRU23609-176
VWR® Traceable® Ultra, Digital Thermometers
Thermometers Digital Thermometers
These thermometers fit into flasks, cuvettes, test tubes and beakers for temperature readings in food, gases, samples, reagents, water baths, ovens, incubators and semi-solids. The long stem allows user to make measurements while stirring solutions in deep vessels. Stainless steel probe (Ø: 3,5 mm).

  • Probe resistant to most chemicals
  • Readings in °C updated every second
  • Safe: Almost unbreakable plastic and stainless steel

Weight: < 28 g

Certifications: Comply with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001.

Delivery information: Supplied with 1More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR