VWR® VistaVision™, Cover Glasses
Supplier: VWR Collection
16004-304 16004-316 16004-308 16004-302 16004-094 16004-326
VWRU16004-304EA 0 INR
VWRU16004-304 VWRU16004-316 VWRU16004-308 VWRU16004-302 VWRU16004-094 VWRU16004-326
VWR® VistaVision™, Cover Glasses
Cover Glasses
Microscope cover glasses made of pure white borosilicate glass with excellent chemical resistance and flatness.
  • Resistant to chemicals
  • Free of bubbles amd scratches
  • No disturbing self-fluorescence under UV light
Certifications: ISO 8255-1
Order Now
SPECIFICATIONS
Environmentally Preferable