VWR®, Biohazard Bags, PP
Supplier: VWR Collection
14220-028 14220-030 14220-032 14220-026
129-0037EA 0 INR
129-0037 VWRU14220-028 VWRU14220-030 VWRU14220-032 VWRU14220-026
VWR®, Biohazard Bags, PP
Bags Waste Disposal Bags
These sturdy bags are ideal for disposing of biologically hazardous waste that must first be autoclaved. PP, 50 µm thick.
  • Autoclavable up to 135 °C
  • Available with or without temperature indicator
  • Useful temperature indicator text is white before autoclaving, then black
  • Text 'Contents Have Been Autoclaved' indicates successful autoclaving
  • Luminous red for easy recognition and highly visible 'Biohazard' label with corresponding warning sign
Bags are formulated to withstand stress at the side seams and are strong enough to hold items in the laboratory.
More Product Information