VWR® Square Media Biotainer Canisters
Supplier: VWR Collection
VWRI210-000110EA 0 INR
VWRI210-000110 VWRI210-000107 VWRI210-000105 VWRI210-000108 VWRI210-000106 VWRI210-000109
VWR® Square Media Biotainer Canisters
Bottles Plastic Bottles
VWR® square containers are ideal for storage of media, laboratory reagents, and biological buffers. Eliminate washing, depyrogenation, and autoclaving steps with sterile and non-pyrogenic containers.
  • Made from durable 100% virgin material
  • Leakproof bottles and HDPE closures
  • Leakproof sealing with integrated lip seal feature
  • Sterile and non-pyrogenic, SAL 10⁻⁶
  • Low extractables
  • Transparency and reduced permeability
All containers are graduated, and feature a moulded hand grip for improved ergonomics.
Polycarbonate containers are suitable for use inMore Product Information