VWR® Classic, Centrifuge Tubes
Supplier: VWR Collection
VWRA525-1557EA 9417.85 INR
VWRA525-1557 VWRA525-1536 VWRA525-1537 VWRA525-1538 VWRA525-1539 VWRA525-1540 VWRA525-1541 VWRA525-1542 VWRA525-1543 VWRA525-1544 VWRA525-1545 VWRA525-1546 VWRA525-1547 VWRA525-1548 VWRA525-1549 VWRA525-1551 VWRA525-1552 VWRA525-1553 VWRA525-1554 VWRA525-1555 VWRA525-1556 VWRA525-1558 VWRA525-1559 VWRA525-1560 VWRA525-1561 VWRA525-1562 VWRA525-1563 VWRA525-1550
VWR® Classic, Centrifuge Tubes
Tubes Centrifuge Tubes
VWR® 15 and 50 ml centrifuge tubes are manufactured in a Class 100000 cleanroom with FDA approved virgin PP.
  • Clear transparent surface
  • Can be centrifuged up to 17000 and 20000 RCF in 15 and 50 ml tube respectively
  • Leak proof screw cap
  • Tamper proof and chemical resistant graduation printing
  • Large writing area for sample identification
  • High quality amber coloured tubes for highly light sensitive sample ensuring no damage to the samples
VWR® classic centrifuge tubes are designed to safeguard precious samples throughout rigorous processes such asMore Product Information