Formamide for use in molecular biology and as a denaturing agent for nucleic acids in gel electrophoresis or hybridization experimen ts 1 * 500 mL
Supplier: VWR Collection
Formamide for use in molecular biology and as a denaturing agent for nucleic acids in gel electrophoresis or hybridization experimen ts 1 * 500 mL
Learn more About VWR
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com