VWR®, DNA Loading Dye Blue, 6x, Ready to Use
Supplier: VWR Collection
VWRA06101100EA 3394.16 INR
VWRA06101100
VWR®, DNA Loading Dye Blue, 6x, Ready to Use
Stains Electrophoresis Stains
VWR® Delivered High quality Reagents & biochemicals, combined with innovative products that gives optimum results , save time & simplify protocols
  • VWR® Lifescience Molecular Biology grade Reagents & Biochemicals
6X DNA Loading dye blue-pre mixed buffer with Bromophenol blue & Xylene cyanol FF .