Safety waste disposal system
Supplier: VITLAB
151594
VITL151594EA 0 INR
VITL151594
Safety waste disposal system
Bottles Plastic Bottles
PE/PP.
  • Good chemical resistance
  • Good temperature resistance
For collection of liquid chemicals in the laboratory. The inlet hopper made from HDPE contains a self-closing float, overfill protection, and a splash guard.
Delivery information: Screw cap (GL 63) with sealing ring is included.
Order Now
SPECIFICATIONS
Environmentally Preferable