Acetic acid glacial_AcOH 99,7 GC, corrected for water content, AR® ACS Trace_metal, Macron Fine Chemicals™

Supplier: Avantor Performance Materials
Danger

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

V193-46EA 0 INR
V193-46 V193-05 MACRV193-18 MACRV193-14 MACRV193-45
Acetic acid glacial_AcOH 99,7 GC, corrected for water content, AR® ACS Trace_metal, Macron Fine Chemicals™
Acetic acid

ACS—Reagents meeting the requirements of the American Chemical Society Committee on Analytical Reagents. The specifications listed in this catalog are based on the ACS Reagent Chemicals Tenth Edition.  

Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 118
Melting Pt: 17
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 38.5
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8 II
REACH: 01-2119475328-30
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements Min 99.7 %
For Laboratory,Research,or Manufacturing Use Max 0.01 %
IMPORTANT: Material will freeze if stored below 17 °C (63°F). Max 0.4 ppm
ACS - Assay (by GC, corrected for water) ≥99.7 %
ACS - Acetic Anhydride ((CH₃CO)₂O) ≤0.01 %
ACS - Chloride (Cl) ≤0.4 ppm
ACS - Color (APHA) ≤10
Dilution Test Passes Test
ACS - Heavy Metals (by ICP-OES) ≤5 ppm
ACS - Residue after Evaporation ≤8 ppm
ACS - Substances Reducing Dichromate Passes Test
ACS - Substances Reducing Permanganate Passes Test
ACS - Sulfate (SO₄) ≤0.4 ppm
ACS - Titrable Base (meq/g) ≤0.0004
Trace Impurities - Aluminum (Al) ≤0.300 ppm
Trace Impurities - Arsenic (As) ≤0.050 ppm
Trace Impurities - Boron (B) ≤0.200 ppm
Trace Impurities - Calcium (Ca) ≤0.300 ppm
Trace Impurities - Chromium (Cr) ≤0.200 ppm
Trace Impurities - Copper (Cu) ≤0.10 ppm
Trace Impurities - Gold (Au) ≤0.300 ppm
Trace Impurities - Iron (Fe) ≤0.2 ppm
Trace Impurities - Lead (Pb) ≤0.300 ppm
Trace Impurities - Magnesium (Mg) ≤0.300 ppm
Trace Impurities - Manganese (Mn) ≤0.300 ppm
Trace Impurities - Nickel (Ni) ≤0.1 ppm
Trace Impurities - Potassium (K) ≤0.300 ppm
Trace Impurities - Sodium (Na) ≤0.300 ppm
Trace Impurities - Tin (Sn) ≤0.300 ppm
Trace Impurities - Titanium (Ti) ≤0.300 ppm
Trace Impurities - Zinc (Zn) ≤0.300 ppm
Appearance (clear, colorless liquid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR