Chemical resistant gloves, uvex rubiflex S XG27B / XG35B
Supplier: UVEX
6056007
UVEX6056007EA 0 INR
UVEX6056007
Chemical resistant gloves, uvex rubiflex S XG27B / XG35B
Gloves
The lightweight safety glove with special NBR (nitrile butadiene rubber) coating and innovative Xtra Grip Technology combines resistance to grease, mineral oils and many chemicals with exceptional comfort and flexibility. The cotton interlock lining is comfortable on the skin and regulates temperature inside the gloves.
  • Ergonomic fit, high flexibility
  • Good resistance to many chemicals
  • Exceptional dry and wet grip
  • Multilayer design for high resistance time
  • High water vapour absorption due to the cotton lining
Ideal for use in the automotive sector, inMore Product Information