Industrial gloves, uvex profi ergo, ENB20A
Supplier: UVEX
6014700 6014701 6014704 6014702 6014703
UVEX6014700EA 0 INR
UVEX6014700 UVEX6014701 UVEX6014704 UVEX6014702 UVEX6014703
Industrial gloves, uvex profi ergo, ENB20A
Gloves
These cotton interlock safety gloves feature a special NBR coating on palm, fingers and ³/₄ of the back of the hand. Ideal for use in oily and greasy areas of application.
  • Very good grip in damp, wet and oily areas
  • Good tactile feel
  • Ergonomic fit, highly flexible
  • Also suitable for operating touch displays
  • Lining provides high wearer comfort, feels pleasant against the skin, and provides optimum regulation of the glove climate
Typical applications include: Assembly, inspection, repair work, shipping/logistics and packaging.
Certifications: EN 388:201More Product Information