Stopcocks, single-way, DURAN®
Supplier: DWK LIFE SCIENCES
286402402 286403407 286403604 286404806 286410805 286411604 286412009 286412403 286412609 286412806 286413408 286413605 286413802 286414601 286414807 286414207 286405408 286406807 286407203 286407409 286407606 286407803 286415006
SCOT286402402EA 0 INR
SCOT286402402 SCOT286403407 SCOT286403604 SCOT286404806 SCOT286410805 SCOT286411604 SCOT286412009 SCOT286412403 201-0383 SCOT286412806 SCOT286413408 SCOT286413605 SCOT286413802 SCOT286414601 SCOT286414807 SCOT286414207 SCOT286405408 SCOT286406807 SCOT286407203 SCOT286407409 SCOT286407606 SCOT286407803 SCOT286415006
Stopcocks, single-way, DURAN®
Stopcocks
DURAN®, borosilicate glass 3.3, clear, with glass or PTFE keys.
  • Replaceable NS keys
  • Screw thread retaining device
Straight bore.
Order Now
SPECIFICATIONS
Environmentally Preferable