Tubing connectors, complete with screw cap, bent, DURAN®
Supplier: DWK LIFE SCIENCES
286222005
SCOT286222005EA 0 INR
SCOT286222005
Tubing connectors, complete with screw cap, bent, DURAN®
Fittings and Connectors
Screw cap PTB, red, with white PP bent hose connection.
  • Very good chemical resistance
  • Very good temperature resistance
With silicone seal (VMQ).