Adapters with standard ground joints, DURAN®
Supplier: DWK LIFE SCIENCES
241142207 241142301 241142404 241142601 241142901 241144405 241142807 241143203 241143306 241143606 241144208 241144302
SCOT241142207EA 0 INR
SCOT241142207 SCOT241142301 SCOT241142404 SCOT241142601 SCOT241142901 SCOT241144405 SCOT241142807 SCOT241143203 201-1704 SCOT241143606 SCOT241144208 SCOT241144302
Adapters with standard ground joints, DURAN®
Adapters Adapters, Conical Ground Joints
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Adapters, reduction or expansion, with standard ground conical joints.
Certifications: DIN 12257
Order Now
SPECIFICATIONS
Environmentally Preferable