Rotating shaker, Rotator Genie™
Supplier: SCIENTIFIC INDUSTRIES ELMIS
SI-2102 SI-2202
SCIISI-2102EA 0 INR
SCIISI-2102 SCIISI-2202
Rotating shaker, Rotator Genie™
Shakers Rotating/Rocking Shakers
These rotators are compact but rugged lab rotators designed to perform for a wide variety of motions which can be adjusted from gentle rolling to forceful end over end mixing. Available in two interchangeable platform configurations, providing the versatility to hold virtually any kind of containers.
  • Ideal for cold rooms and incubators (will easily fit in most incubators)
  • Standard platform with retaining bands is designed to turn 90 degrees allowing different mixing motions
  • Stainless steel magnetic platform features unique spring clip plate system