Orbital shakers, Orbital-Genie™
Supplier: SCIENTIFIC INDUSTRIES ELMIS
SI-1502 SI-1503 SI-1504 SI-1602 SI-1603 SI-1604 SI-1702 SI-1703 SI-1704
SCIISI-1502EA 0 INR
SCIISI-1502 SCIISI-1503 SCIISI-1504 SCIISI-1602 SCIISI-1603 SCIISI-1604 SCIISI-1702 SCIISI-1703 SCIISI-1704
Orbital shakers, Orbital-Genie™
Shakers Orbital Shakers
Large platform shaker with choice of different platform options: Universal ratcheting clamp, adhering mat or unit with standard
flask clamps.
  • Variable shaking speed with orbit reversing
  • Fully programmable - time, reversing time, acceleration/deceleration rates, and variable run/stop time
  • Create any mixing action from a smooth vortex to sloshing aeration (or anything in between)
  • Equipped with stepper motor, delivering exceptional speed control, accuracy, and durability
  • Timer displays time or, when programmed to a defined limit, will shut off the unit
  • Can be
More Product Information