Refrigerated shaking incubators, multi-functional, Incubator-Genie™ with accessories
Supplier: SCIENTIFIC INDUSTRIES ELMIS
SI-1402 SI-1403 SI-1404 SI-1401
SCIISI-1402EA 0 INR
SCIISI-1402 SCIISI-1403 SCIISI-1404 SCIISI-1401
Refrigerated shaking incubators, multi-functional, Incubator-Genie™ with accessories
Shakers Shaking Incubators
Allows to heat, rotate, rock, and shake all in one compact unit.
  • Gently and continuously recirculated air ensures temperature uniformity
  • Viewing window to inspect samples without affecting chamber environment
  • Magnetic rotating/rocking: Platform clip plate system holds almost any container at any angle
  • Compatible with an extensive accessory line-up
The incubator is best for applications such as Gel & Blot staining, Cell or Bacterial Culture, as well as any application that requires mixing, washing, rocking, and temperature control of a sample.