Sodium thiosulphate pentahydrate AR, Rankem®

Supplier: RANKEM

S0439
RANKS0439EA 0 INR
RANKS0439
Sodium thiosulphate pentahydrate AR, Rankem®
Sodium thiosulphate pentahydrate
Formula: Na₂S₂O₃.5H₂O
MW: 248.19 g/mol
Melting Pt: 48
MDL Number: MFCD00149186
CAS Number: 10102-17-7
Merck Index: 12,08844

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR