Perchloric acid solution_SC 0,1 N, Analytical reagent, Rankem®

Supplier: RANKEM
Warning

P0049
RANKP0049EA 2105.4 INR
RANKP0049
Perchloric acid solution_SC 0,1 N, Analytical reagent, Rankem®
Perchloric acid

High purity analytical reagent.


  • Ready to use solution for 0,1 N


Description: A clear, colourless liquid with a pungent odour.

Formula: HClO₄
MW: 100.46 g/mol
Boiling Pt: 39
Melting Pt: -101
MDL Number: MFCD00011325
CAS Number: 7601-90-3
EINECS: 231-512-4
UN: 3264
ADR: 8 III

Order Now

Specification Test Results

Normality 0.095 - 0.105 N
Water ≤ 0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR