Rankem®, Secondary Reference Standards for GC
Supplier: RANKEM
GCI001-100 GCT001-100 GCD001-100 GCM001-100 GCI003-100 GCM003-100 GCN001-100 GCD002-100 GCA001-100 GCC001-100 GCD003-100 GCN002-100 GCN003-100 GCE001-100 GCM004-100 GCT002-100 GCA002-100 GCN004-100 GCE002-100 GCO001-100 GCC002-100 GCN005-100 GCN006-100 GCB001-100 GCD004-100 GCT003-100 GCI004-100 GCD005-100 GCP002-100 GCM005-100 GCA003-100 GCD006-100 GCB002-100 GCP003-100 GCM006-100 GCE003-100 GCE004-100 GCD007-100 GCI005-100 GCB003-100 GCP004-100 GCD008-100 GCA004-100 GCM007-100 GCT004-100 GCB004-100 GCN007-100 GCI006-100 GCB005-100 GCN008-100 GCN009-100 GCC004-100 GCC005-100 GCP005-100 GCD009-100 GCM008-100 GCP006-100 GCD010-100 GCD011-100 GCC006-100 GCB006-100 GCN010-100 GCD012-100 GCT005-100 GCP007-100 GCM009-100 GCT006-100 GCC007-100 GCA005-100 GCD013-100 GCD014-100 GCP008-100 GCE005-100 GCO002-100 GCS001-100 GCI007-100 GCM010-100 GCM011-100 GCT007-100 GCM012-100 GCE006-100 GCM013-100 GCH001-100 GCF001-100 GCC008-100 GCM014-100 GCM015-100 GCP009-100 GCC009-020 GCN012-100 GCC010-100 GCC011-100 GCP010-100 GCE008-100 GCB009-100 GCD016-100 GCN013-100 GCB010-100 GCP011-100 GCI010-100 GCT008-100 GCD017-100 GCT009-100 GCD018-100 GCM016-100 GCB011-100 GCE009-100 GCM017-100 GCC012-100 GCN014-100 GCP013-100 GCE010-100 GCP014-100 GCP015-100 GCC013-100 GCD019-100 GCD020-100 GCT010-100 GCT011-100 GCE011-100 GCB012-100 GCC014-100 GCC015-100 GCT012-100 GCD021-100 GCT013-100 GCC016-100 GCC017-100 GCM018-100 GCB013-100 GCT014-100 GCD022-100 GCI011-100 GCD023-100 GCB014-100 GCD024-100 GCD025-100 GCA006-100 GCT015-100 GCD026-100 GCX001-100 GCC018-100 GCH002-100 GCI012-100 GCN015-100 GCM019-100 GCD027-100 GCA007-100 GCT016-100 GCD028-100 GCD029-100 GCD030-100 GCT017-100 GCP016-100 GCP017-100 GCP018-100 GCP019-100 GCD031-100 GCI013-100 GCO003-100 GCM020-100 GCP020-100 GCS002-100 GCT018-100 GCN016-100 GCP021-100 GCI014-100 GCN017-100 GCT019-100 GCC019-100 GCR001-100 GCN018-100 GCA008-100 GCM022-100 GCP022-100 GCO005-100
RANKGCI001-100EA 10731.64 INR
RANKGCI001-100 RANKGCT001-100 RANKGCD001-100 RANKGCM001-100 RANKGCI003-100 RANKGCM003-100 RANKGCN001-100 RANKGCD002-100 RANKGCA001-100 RANKGCC001-100 RANKGCD003-100 RANKGCN002-100 RANKGCN003-100 RANKGCE001-100 RANKGCM004-100 RANKGCT002-100 RANKGCA002-100 RANKGCN004-100 RANKGCE002-100 RANKGCO001-100 RANKGCC002-100 RANKGCN005-100 RANKGCN006-100 RANKGCB001-100 RANKGCD004-100 RANKGCT003-100 RANKGCI004-100 RANKGCD005-100 RANKGCP002-100 RANKGCM005-100 RANKGCA003-100 RANKGCD006-100 RANKGCB002-100 RANKGCP003-100 RANKGCM006-100 RANKGCE003-100 RANKGCE004-100 RANKGCD007-100 RANKGCI005-100 RANKGCB003-100 RANKGCP004-100 RANKGCD008-100 RANKGCA004-100 RANKGCM007-100 RANKGCT004-100 RANKGCB004-100 RANKGCN007-100 RANKGCI006-100 RANKGCB005-100 RANKGCN008-100 RANKGCN009-100 RANKGCC004-100 RANKGCC005-100 RANKGCP005-100 RANKGCD009-100 RANKGCM008-100 RANKGCP006-100 RANKGCD010-100 RANKGCD011-100 RANKGCC006-100 RANKGCB006-100 RANKGCN010-100 RANKGCD012-100 RANKGCT005-100 RANKGCP007-100 RANKGCM009-100 RANKGCT006-100 RANKGCC007-100 RANKGCA005-100 RANKGCD013-100 RANKGCD014-100 RANKGCP008-100 RANKGCE005-100 RANKGCO002-100 RANKGCS001-100 RANKGCI007-100 RANKGCM010-100 RANKGCM011-100 RANKGCT007-100 RANKGCM012-100 RANKGCE006-100 RANKGCM013-100 RANKGCH001-100 RANKGCF001-100 RANKGCC008-100 RANKGCM014-100 RANKGCM015-100 RANKGCP009-100 RANKGCC009-020 RANKGCN012-100 RANKGCC010-100 RANKGCC011-100 RANKGCP010-100 RANKGCE008-100 RANKGCB009-100 RANKGCD016-100 RANKGCN013-100 RANKGCB010-100 RANKGCP011-100 RANKGCI010-100 RANKGCT008-100 RANKGCD017-100 RANKGCT009-100 RANKGCD018-100 RANKGCM016-100 RANKGCB011-100 RANKGCE009-100 RANKGCM017-100 RANKGCC012-100 RANKGCN014-100 RANKGCP013-100 RANKGCE010-100 RANKGCP014-100 RANKGCP015-100 RANKGCC013-100 RANKGCD019-100 RANKGCD020-100 RANKGCT010-100 RANKGCT011-100 RANKGCE011-100 RANKGCB012-100 RANKGCC014-100 RANKGCC015-100 RANKGCT012-100 RANKGCD021-100 RANKGCT013-100 RANKGCC016-100 RANKGCC017-100 RANKGCM018-100 RANKGCB013-100 RANKGCT014-100 RANKGCD022-100 RANKGCI011-100 RANKGCD023-100 RANKGCB014-100 RANKGCD024-100 RANKGCD025-100 RANKGCA006-100 RANKGCT015-100 RANKGCD026-100 RANKGCX001-100 RANKGCC018-100 RANKGCH002-100 RANKGCI012-100 RANKGCN015-100 RANKGCM019-100 RANKGCD027-100 RANKGCA007-100 RANKGCT016-100 RANKGCD028-100 RANKGCD029-100 RANKGCD030-100 RANKGCT017-100 RANKGCP016-100 RANKGCP017-100 RANKGCP018-100 RANKGCP019-100 RANKGCD031-100 RANKGCI013-100 RANKGCO003-100 RANKGCM020-100 RANKGCP020-100 RANKGCS002-100 RANKGCT018-100 RANKGCN016-100 RANKGCP021-100 RANKGCI014-100 RANKGCN017-100 RANKGCT019-100 RANKGCC019-100 RANKGCR001-100 RANKGCN018-100 RANKGCA008-100 RANKGCM022-100 RANKGCP022-100 RANKGCO005-100
Rankem®, Secondary Reference Standards for GC
Standards Chromatography Standards
Rankem offers a wide range of highly pure & characterised secondary GC reference standards for chromatographic and analytical applications.
  • A wide range of highly pure reference substances for GC (>170 std)
  • Free from inorganic non volatile impurities by analysing residue after evaporation.
  • The reference substances are supplied in screw-capped amber coloured glass bottles
Rankem GC reference standards have typically over 99% purity by GC-FID. Identifications determined by GC-MS, FTIR and 1H NMR are reported in the certificate of analysis.
Delivery information: Each pack of GC reference standard comes with a gas chromatogram generated under appropriate testing conditions, along with the structures confirmed by Spectral analysis such as Mass, IR and NMR