n-Hexadecane, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Hexadecane, Cetane

N105-07EA 0 INR
N105-07
n-Hexadecane, J.T.Baker®
n-Hexadecane
Formula: H₃C(CH₂)₁₄CH₃
MW: 226.45 g/mol
Boiling Pt: 287
Melting Pt: 18
Density: 0.77331 g/cm³ (20 °C)
Flash Pt: 135 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008998
CAS Number: 544-76-3
EINECS: 208-878-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90d3084bdd908b305ecf9b3c30e03cd98a407d94&vid=e5a15f4f6c36024a34521e08083957252ff78457&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Identification (by IR) Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR