CL-1000i, Fully automatic Chemiluminescence Immunoassay (CLIA) System
Supplier: SHENZHEN MINDRAY BIO MEDICAL
BM20B-PA00005
MINDBM20B-PA00005EA 2236320.29 INR
MINDBM20B-PA00005
CL-1000i, Fully automatic Chemiluminescence Immunoassay (CLIA) System
Clinical Diagnostic Supplies Clinical Analysers
CL-1000i, is a robust and simple to use chemiluminescence analyzer with a throughput of up to 120 tests per hour. In a compact benchtop package, it has a large onboard capacity of 25 reagents and 60 samples, and supports non-stop refill of samples, reagents and consumables during testing. By integrating the reliable CLIA technology with latest electrical and software improvement, CL-1000i enables easy operation and brings confident diagnosis faster than ever.
  • 120 Tests/hr
  • 25 Positions for Reagents
  • 60 Samples positions
  • Ready to Use Wash Buffers