Chemfluor® Smooth PTFE-Lined Hose Assemblies
Supplier: Avantor Fluid Handling
MFLX96552-01EA 16200 INR
MFLX96552-01 MFLX96552-12 MFLX96553-01 MFLX96553-12 MFLX96553-13 MFLX96555-02
Chemfluor® Smooth PTFE-Lined Hose Assemblies
Tubing Transfer Tubing
Meets the demands of various industries.
  • Ideal for general-purpose, single-use, and extended fluid path applications
Applications: Sanitary or steam transfer, bottle filling, gas analysis, hydraulic lines, extrusion presses, molding and adhesives
Construction
Tube: Chemfluor® PTFE
Reinforcement: 304 stainless steel braid
Advantages: Thicker PTFE tube wall. Superior kink resistance. Excellent vacuum rating. Neutral to taste, color and odor. Nonstick and noncontaminating. Easily cleaned.
Fittings: Permaseal® 316L stainless steel hex male, female JIC, hexMore Product Information