Masterflex® I/P® Precision Pump Tubing, Platinum-Cured Silicone, Avantor®
Supplier: Avantor Fluid Handling
MFLX96410-26EA 48887.12 INR
MFLX96410-26 MFLX96410-73 MFLX96410-82
Masterflex® I/P® Precision Pump Tubing, Platinum-Cured Silicone, Avantor®
Tubing Peristaltic Pump Tubing
Reduce the risk of particulate entrapment with smooth inner surface.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory Approvals to match your unique application
  • No leachable additives protects sensitive and
More Product Information