Masterflex® Transfer Tubing, Tygon® 2475 I.B., High Purity Reinforced, Avantor®

Supplier: Avantor Fluid Handling
95360-50 95360-51 95360-52 95360-53 95360-55 95360-54
MFLX95360-50EA 66000 INR
MFLX95360-50 MFLX95360-51 MFLX95360-52 MFLX95360-53 MFLX95360-55 MFLX95360-54
Masterflex® Transfer Tubing, Tygon® 2475 I.B., High Purity Reinforced, Avantor®
Tubing Transfer Tubing
Keeps visual track of samples.

  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application

Formulation: TYGON® 2475 I.B.
Applications: Pharmaceutical and cosmetic processing, deionized water systems, cell harvest and media process systems, chemical transfer, preservative and sterile fill lines.
Characteristics: PlasticizerMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR