Masterflex® Transfer Tubing, PVDF, Avantor®
Supplier: Avantor Fluid Handling
95100-04
MFLX95100-00EA 17670.59 INR
MFLX95100-00 MFLX95100-02 MFLX95100-06 MFLX95100-10 MFLX95100-12 MFLX95100-14 MFLX95100-16 MFLX95100-04
Masterflex® Transfer Tubing, PVDF, Avantor®
Tubing Transfer Tubing
Fluoropolymer tubing at less cost.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
This tubing is ideal for high-purity applications and corrosive chemical transfer—an inexpensive alternative to fluoropolymer tubing. The polyvinylidene fluoride (PVDF) material is kink resistant, has excellent chemicalMore Product Information