Masterflex® 8 Place Centrifuge Rotor, Avantor®
Supplier: Avantor Fluid Handling
83058-04
MFLX83058-04EA 0 INR
MFLX83058-04
Masterflex® 8 Place Centrifuge Rotor, Avantor®
Centrifuges Rotors
Compatible with Cole-Parmer® FS-3400 and VS-4000 Centrifuges.
This optional rotor is compatible with Cole-Parmer® centrifuges, models 83058-00, -02, and -42. This rotor will hold 8×10 ml tubes. Optional tube adapters are sold separately as part numbers 83058-08 to 4 to 5 ml tube and 83058-10 for 2 ml tubes. Centrifuge is sold separately.