Remote-Control Diaphragm Liquid Dosing Pumps
Supplier: Avantor Fluid Handling
78167-09 78167-00 78167-01 78167-02 78167-05 78167-07 78167-10
MFLX78167-09EA 514000 INR
MFLX78167-09 MFLX78167-00 MFLX78167-01 MFLX78167-02 MFLX78167-05 MFLX78167-07 MFLX78167-10
Remote-Control Diaphragm Liquid Dosing Pumps
Pumps Diaphragm Pumps
Uniformity of discharge ensures low-pulsation transfer process.
  • Quiet, reliable, accurate, and virtually maintenance-free
  • Pumps can run dry continuously without damage
  • Versatile pumps handle flow rates up to 100 ml/min and volumes up to 1000 mL
Simple and intuitive function makes the liquid dosing pump an accurate and reliable solution for your laboratory dosing applications. User-friendly design provides a quiet, dry-running, and self-priming dosing operation. Easy calibration and cycle mode allow for uninterrupted and virtually maintenance-freeMore Product Information