Masterflex® Ismatec® Reglo Digital Miniflex Pumps, Avantor®
Supplier: Avantor Fluid Handling
MFLX78018-40EA 421000 INR
MFLX78018-40 MFLX78018-42 MFLX78018-40-IN MFLX78018-42-IN MFLX78018-40-IL MFLX78018-42-IL MFLX78018-40-CH MFLX78018-42-CH
Masterflex® Ismatec® Reglo Digital Miniflex Pumps, Avantor®
Pumps Peristaltic Pumps
Versatile pumps for accurate low-flow fluid delivery – with an intuitive touch-screen interface.
  • Compact, benchtop pumps optimize ease of use, accuracy, and reliability
  • 5” (12.7 cm) capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
  • Flexible operational control via screen or analog remote control
  • Continuous run, volume dispense, and time dispense modes -- batch control and interval/off time in Volume and Time modes
  • Pump lets you save and retrieve commonly used dispense programs for rapid set up
  • Anti-drip and speed ramping
More Product Information