Masterflex® Ismatec® Reglo Independent Channel Control (ICC) Peristaltic Pumps, Avantor®
Supplier: Avantor Fluid Handling
MFLX78001-60EA 475000 INR
MFLX78001-60 MFLX78001-68 MFLX78001-70 MFLX78001-80 MFLX78001-82 MFLX78001-58-IN MFLX78001-60-IN MFLX78001-62-IN MFLX78001-68-IN MFLX78001-70-IN MFLX78001-72-IN MFLX78001-78-IN MFLX78001-80-IN MFLX78001-82-IN MFLX78001-58-IL MFLX78001-60-IL MFLX78001-62-IL MFLX78001-68-IL MFLX78001-70-IL MFLX78001-72-IL MFLX78001-78-IL MFLX78001-80-IL MFLX78001-82-IL MFLX78001-58-CH MFLX78001-60-CH MFLX78001-62-CH MFLX78001-68-CH MFLX78001-70-CH MFLX78001-72-CH MFLX78001-78-CH MFLX78001-80-CH MFLX78001-82-CH
Masterflex® Ismatec® Reglo Independent Channel Control (ICC) Peristaltic Pumps, Avantor®
Pumps Peristaltic Pumps
Independently control flow and direction of each channel via keypad or your PC.
  • Flow rate 0.001 to 43 ml/min per channel - flow rate depends on drive rpm, rollers, and tubing size
  • USB interface for quick connection to a PC
  • Flexibility of bidirectional flow in each channel
These multi-channel peristaltic pumps let you individually control the flow and direction of each channel from the intuitive keypad or via your computer—expanding your benchtop pumping capabilities with a single compact unit! Pump delivers continuous pumping or precision dispensingMore Product Information