Masterflex® Ismatec® IPC and IPC-N Digital Multichannel Peristaltic Pumps, Avantor®
Supplier: Avantor Fluid Handling
78000-03 78000-11 78000-13 78000-23 78000-31 78000-41 78001-00 78001-02 78001-10 78001-12 78001-20 78001-22 78001-32 78001-40 78001-42 78000-43
MFLA78000-03EA 683000 INR
MFLA78000-03 MFLA78000-11 MFLA78000-13 MFLA78000-23 MFLA78000-31 MFLA78000-41 MFLA78001-00 MFLA78001-02 MFLA78001-10 MFLA78001-12 MFLA78001-20 MFLA78001-22 MFLA78001-32 MFLA78001-40 MFLA78001-42 MFLX78000-09-IN MFLX78000-19-IN MFLX78000-29-IN MFLX78000-37-IN MFLX78000-39-IN MFLX78000-47-IN MFLX78000-49-IN MFLX78001-06-IN MFLX78001-08-IN MFLX78001-16-IN MFLX78001-18-IN MFLX78001-26-IN MFLX78001-28-IN MFLX78001-36-IN MFLX78001-38-IN MFLX78001-46-IN MFLX78001-48-IN MFLX78000-09-IL MFLX78000-19-IL MFLX78000-29-IL MFLX78000-37-IL MFLX78000-39-IL MFLX78000-47-IL MFLX78000-49-IL MFLX78001-06-IL MFLX78001-08-IL MFLX78001-16-IL MFLX78001-18-IL MFLX78001-26-IL MFLX78001-28-IL MFLX78001-36-IL MFLX78001-38-IL MFLX78001-46-IL MFLX78001-48-IL MFLX78000-09-CH MFLX78000-19-CH MFLX78000-29-CH MFLX78000-37-CH MFLX78000-39-CH MFLX78000-47-CH MFLX78000-49-CH MFLX78001-06-CH MFLX78001-08-CH MFLX78001-16-CH MFLX78001-18-CH MFLX78001-26-CH MFLX78001-28-CH MFLX78001-36-CH MFLX78001-38-CH MFLX78001-46-CH MFLX78001-48-CH MFLX78000-43 MFLX78000-17-IN MFLX78000-43-USS MFLX78000-17-IL
Masterflex® Ismatec® IPC and IPC-N Digital Multichannel Peristaltic Pumps, Avantor®
Pumps Peristaltic Pumps
Fewer tubing changes!
  • Planetary drive with 8 stainless steel rotors reduces pulsation, increases flow rate accuracy, and lengthens tubing life
  • Control via 6-button keypad, computer, or remote control
  • Four-digit LED display facilitates repeatability
  • Automatic occlusion cartridges are easy to use and provide reproducible results
These Ismatec® IPC and IPC-N digital peristaltic multichannel programmable pumps feature planetary gears that drive eight stainless steel rollers in unison for smooth rotation and accurate liquid flow. The four-digit LED displayMore Product Information