Masterflex® L/S® MasterSense™ Fill/Finish Pump System with Masterflex Ultrapharm™ Pump Head, Avantor®
Supplier: Avantor Fluid Handling
77976-40
MFLX77976-40EA 2450548.01 INR
MFLX77976-40 MFLX77976-40-IN MFLX77976-42-IN MFLX77976-40-IL MFLX77976-42-IL
Masterflex® L/S® MasterSense™ Fill/Finish Pump System with Masterflex Ultrapharm™ Pump Head, Avantor®
Pumps Peristaltic Pumps
A highly accurate benchtop pump system for dosing, dispensing, and fill/finish applications.
  • Offset rollers and planetary gearing ensure dosing accuracy (±0,5%), low pulsation, and long tubing life
  • Masterflex double-Y tube sets ensure proper retention, repeatability, and accuracy
  • 250 rpm max speed; dosing volumes from <0,5 to >500 ml
  • Easy on tubing—planetary drive rollers reduce drag on tubing for longer tubing life and reduced risk of premature tubing failure
  • Whisper quiet operation reduces extra noise on the floor or in the lab
  • Autoclavable pump
More Product Information