Masterflex® I/P® Digital Process Drives, Avantor®
Supplier: Avantor Fluid Handling
77420-20 77420-40
MFLX77420-10EA 1016208.05 INR
MFLX77420-10 MFLX77420-20 MFLA77420-40 MFLX77420-10-IN MFLX77420-18-IN MFLX77420-20-IN MFLX77420-28-IN MFLX77420-30-IN MFLX77420-32-IN MFLX77420-40-IN MFLX77420-42-IN MFLX77420-10-IL MFLX77420-18-IL MFLX77420-20-IL MFLX77420-28-IL MFLX77420-30-IL MFLX77420-32-IL MFLX77420-40-IL MFLX77420-42-IL MFLX77420-10-CH MFLX77420-18-CH MFLX77420-20-CH MFLX77420-28-CH MFLX77420-30-CH MFLX77420-32-CH MFLX77420-40-CH MFLX77420-42-CH
Masterflex® I/P® Digital Process Drives, Avantor®
Pumps Pump Drives Peristaltic Pump Drives
Maintenance-free drives built for precision dispensing.
  • Flow rate 0.0006 to 26 LPM with I/P tubing—flow rate depends on drive rpm, pump head, number of pump heads, and tubing size
  • Digital interface with graphical LCD shows pump performance and operating mode— continuous run, timed dispense, copy dispense, or volume dispense
  • Remote control available using analog communication—voltage, current and contact closure
  • Select your preferred language from the user-friendly menu for programming and operation
  • Dispense by Volume in ml (0.001 to 9,999 mL), L, or gal;
More Product Information