Masterflex® L/S® Easy-Load® II Pump Heads for High-Performance Precision Tubing, Avantor®
Supplier: Avantor Fluid Handling
77200-62
MFLX77200-32EA 75331.03 INR
MFLX77200-32 MFLX77200-52 MFLX77200-62 MFLX77201-62
Masterflex® L/S® Easy-Load® II Pump Heads for High-Performance Precision Tubing, Avantor®
Pumps Pump Heads Peristaltic Pump Heads
Automatic retention holds tubing securely in place with no manual adjustment—just load the tubing and start the pump.
  • New design and automatic tubing retention allow for multiple tubing sizes—change tubing without removing head from drive
  • Four-roller rotor improves pressure performance and mechanical stability while reducing pulsation
  • Model available with integrated open-head sensor–connects directly to sensor-equipped drives
  • Pump heads are stackable for multichannel applications
  • Compact design for efficient use of space
  • Suitable for use with L/S® High-
More Product Information