ITT Jabsco Sanitary Wash-Down Rotary Lobe Pumps
Supplier: Avantor Fluid Handling
75201-00 75201-10
MFLX75201-00EA 2305000 INR
MFLX75201-00 MFLX75201-10
ITT Jabsco Sanitary Wash-Down Rotary Lobe Pumps
Pumps Rotary Lobe Pumps
Low maintenance rotary lobe pumps conform to 3A sanitary standards.
  • Front mounted shaft seals are fully accessible from the front of the pump for easy replacement without disturbing piping in the process line.
  • Bi-wing rotors require no timing adjustments and provide a high volumetric efficiency.
  • Rugged design incorporates heavy-duty shafts for high-pressure capability.
Pumps are approved to EHEDG (European Hygienic Equipment Design Group) and CIP/SIP standards. Remove pump casing for quick maintenance and cleaning. Reduced noise and cavitation provideMore Product Information