Neptune Hydraulically Actuated Diaphragm Pumps
Supplier: Avantor Fluid Handling
74151-30 74151-00 74151-02 74151-10 74151-22 74151-40
MFLX74151-30EA 515000 INR
MFLX74151-30 MFLX74151-00 MFLX74151-02 MFLX74151-10 MFLX74151-22 MFLX74151-40
Neptune Hydraulically Actuated Diaphragm Pumps
Pumps Diaphragm Pumps
High suction lift and high viscosity combine in the stainless steel pump.
  • Achieve up to 65 GPH or 1100 psi
  • Internal bypass allows for improved valve performance
  • Pump capacity is adjustable through 100% of range using a manual micrometer dial/automatic electric positioner with manual override
Pump drive components are fully enclosed and operate in an oil bath to minimize friction and wear, as well as to protect from harsh environments. Pump provides metering accuracy of ±1%. A micrometer knob enables capacity adjustment between 0 to 100% whether theMore Product Information