Walchem High Stroke-Frequency Diaphragm Metering Pumps
Supplier: Avantor Fluid Handling
74126-00 74126-10 74126-20 74126-30 74126-35 74127-22 74125-00 74127-30
MFLX74126-00EA 95500 INR
MFLX74126-00 MFLX74126-10 MFLX74126-20 MFLX74126-30 MFLX74126-35 MFLX74127-22 MFLX74125-00 MFLX74127-30
Walchem High Stroke-Frequency Diaphragm Metering Pumps
Pumps Diaphragm Pumps
Small pump cavity, two check valves, and high stroke frequency combine for superb self-priming and degassing properties.
  • Pumps provide up to 360 strokes per minute for smooth, efficient operation
  • Pumps prime within seconds of being turned on; prime is maintained by two check valves (one on each side)
  • Pump housing is IP65 rated
These Walchem high stroke-frequency diaphragm metering pumps provide smooth operation combined with self-priming capabilities within seconds. These pumps are available in three options—choose from manual control, remoteMore Product Information