Rechargeable Electric Backpack Sprayer
Supplier: Avantor Fluid Handling
70605-05
MFLX70605-05EA 123000 INR
MFLX70605-05
Rechargeable Electric Backpack Sprayer
Spray Guns
Microprocessor pump control with adjustable spray tip – delicate mist, high volume, or long-range stream.
  • Tank capacity of 4 gallons
  • Comfortable ergonomic design with a full back pad and thickly padded shoulder harness.
  • Battery pack charges while in place or can be removed for remote charging.
Lightweight polyethylene tank is engineered to match the body's contour. Hand lever provides on-demand operation — depress to begin spraying and release to stop. Hand lever may also be locked in the open position to reduce operator fatigue during longerMore Product Information