Basic Variable-Speed DC Motor Drives
Supplier: Avantor Fluid Handling
70100-10
MFLX70100-10EA 100000 INR
MFLX70100-10
Basic Variable-Speed DC Motor Drives
Pumps Motors
Vary the speed of your DC motor with these controllers.
  • Easily control the speed of your DC motor
Basic Controller keeps 1/4- to 2-hp motors at constant speeds without additional resistors. The NEMA 1 (IP 10) housing guards against dust, light, and incidental splashing.
For thermal overload protection, use heat sink model 70100-50 with 90 VDC motors (3/4 hp or larger) and 180 VDC motors (1-1/2 hp or larger)—order separately.
Delivery information: Includes instruction operation manual.