Masterflex® Quick-Disconnect Fittings, Avantor®
Supplier: Avantor Fluid Handling
70000-37 70000-38 70000-31 70000-33 70000-34 70000-44
MFLX70000-37EA 1130.91 INR
MFLX70000-37 MFLX70000-38 MFLX70000-31 MFLX70000-33 MFLX70000-34 MFLX70000-44 MFLX70000-24 MFLX70000-25 MFLX70000-26 MFLX70000-28 MFLX70000-29
Masterflex® Quick-Disconnect Fittings, Avantor®
Fittings and Connectors
Disconnect and reconnect your flow path with ease.
  • Audible click lets you know the coupling is secure
  • Many hose barb sizes to choose from
  • Valved design allows you to cut off fluid flow when the fittings are disconnected
Use these fittings to quickly connect and disconnect from your flow line. Simply order a body (female half) and an insert (male half). Couplings are compatible for use with gases and liquids. Choose between acetal for a wide operating temperature range and greater durability or polypropylene (PP) for stronger chemical resistance andMore Product Information